Anti-ADD1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB99354-20EA 0 CHF
BIRBORB99354-20 BIRBORB99354-100
Anti-ADD1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: ADD1 (adducin 1 (alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR