Anti-SHC1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB99350-100EA 0 CHF
BIRBORB99350-100 BIRBORB99350-20
Anti-SHC1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR