Anti-NR3C1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB99323-20EA 0 CHF
BIRBORB99323-20 BIRBORB99323-100
Anti-NR3C1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: NR3C1 (nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR