Anti-F7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40823-100EA 424 CHF
BIRBORB40823-100
Anti-F7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: F7 (coagulation factor VII (serum prothrombin conversion accelerator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR