Anti-HAUS8 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40807-100EA 424 CHF
BIRBORB40807-100
Anti-HAUS8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: HAUS8 (HAUS augmin-like complex, subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR