Anti-APEX1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40741-100EA 424 CHF
BIRBORB40741-100
Anti-APEX1 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR