Anti-IgA Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27697-0.5EA 0 CHF
BIRBORB27697-0.5 BIRBORB27697-0.25
Anti-IgA Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR