Anti-CHEK1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27666-100EA 0 CHF
BIRBORB27666-100
Anti-CHEK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR