Anti-FIGF Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27547-100EA 616 CHF
BIRBORB27547-100
Anti-FIGF Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: FIGF (c-fos induced growth factor (vascular endothelial growth factor D))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR