Anti-p170 Glycoprotein Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27448-1EA 0 CHF
BIRBORB27448-1
Anti-p170 Glycoprotein Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: p170 Glycoprotein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR