Anti-INS Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27420-1EA 0 CHF
BIRBORB27420-1
Anti-INS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: INS (insulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR