Anti-Laminin 5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26824-10EA 0 CHF
BIRBORB26824-10
Anti-Laminin 5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Laminin 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR