Anti-Mouse CD45 (LCA) Rat Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26410-0.5EA 0 CHF
BIRBORB26410-0.5
Anti-Mouse CD45 (LCA) Rat Monoclonal Antibody
Antibodies
Flow Cytometry: 1 ?g

Type: Primary
Antigen: Mouse CD45 (LCA)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b (Rat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR