Anti-AFP Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26139-1EA 697 CHF
BIRBORB26139-1
Anti-AFP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR