Anti-CKB Mouse monoclonal antibody

Lieferant: Biorbyt
BIRBORB26046-1EA 607 CHF
BIRBORB26046-1
Anti-CKB Mouse monoclonal antibody
Antikörper
Type: Primary
Antigen: CKB (creatine kinase, brain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR