Anti-PSA, Total Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB26026-1EA 1259 CHF
BIRBORB26026-1
Anti-PSA, Total Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: PSA, Total
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR