Anti-PSA, Total Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26020-1EA 783 CHF
BIRBORB26020-1
Anti-PSA, Total Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PSA, Total
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR