Anti-UMOD Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25734-1EA 0 CHF
BIRBORB25734-1
Anti-UMOD Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UMOD (uromodulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR