Anti-CD37 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB25700-100EA 0 CHF
BIRBORB25700-100
Anti-CD37 Mouse Monoclonal Antibody
Antibodies
  • ELISA: 1:1000

Type: Primary
Antigen: CD37 (CD37 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR