Anti-CCR3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25684-100EA 424 CHF
BIRBORB25684-100
Anti-CCR3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1-2 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CCR3 (chemokine (C-C motif) receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR