Anti-Avidin, egg white Mouse Monoclonal Antibody
Supplier: Biorbyt
BIRBORB25584-1EA 624 CHF
BIRBORB25584-1
Anti-Avidin, egg white Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Avidin, egg white
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now