Anti-APOE Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB25254-0.5EA 1216 CHF
BIRBORB25254-0.5
Anti-APOE Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
DB: 1:200-1:2000; ELISA: 1:2000-1:8000

Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR