Anti-FTH1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25252-1EA 0 CHF
BIRBORB25252-1
Anti-FTH1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR