Anti-XPO7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB25124-100EA 424 CHF
BIRBORB25124-100
Anti-XPO7 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 1:500

Type: Primary
Antigen: XPO7 (exportin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR