Anti-Myoglobin Cardiac Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24976-1EA 1108 CHF
BIRBORB24976-1
Anti-Myoglobin Cardiac Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Myoglobin Cardiac
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR