Anti-pro ANP Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24706-10EA 0 CHF
BIRBORB24706-10
Anti-pro ANP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: pro ANP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR