Anti-Hemoglobin A1c Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24670-1EA 857 CHF
BIRBORB24670-1
Anti-Hemoglobin A1c Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Hemoglobin A1c
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR