Anti-ALB Goat Polyclonal Antibody (Biotin)
Supplier: Biorbyt
BIRBORB24603-2EA 0 CHF
BIRBORB24603-2
Anti-ALB Goat Polyclonal Antibody (Biotin)
Antibodies
ELISA: 1:20000
Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Human