Anti-Procalcitonin/Katacalcin Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24414-1EA 1236 CHF
BIRBORB24414-1
Anti-Procalcitonin/Katacalcin Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Procalcitonin/Katacalcin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR