Anti-CMV pp65 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24139-100EA 0 CHF
BIRBORB24139-100
Anti-CMV pp65 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CMV pp65
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR