Anti-B. anthracis SA Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB24082-1EA 1182 CHF
BIRBORB24082-1
Anti-B. anthracis SA Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: B. anthracis SA (Bacillus anthracis spore antigen)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR