Anti-HBsAg (ayw) Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB24066-1EA 656 CHF
BIRBORB24066-1
Anti-HBsAg (ayw) Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: HBsAg (ayw)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR