Anti-Adenovirus Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB23922-1EA 697 CHF
BIRBORB23922-1
Anti-Adenovirus Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Adenovirus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR