Anti-Adenovirus hexon Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB23776-100EA 0 CHF
BIRBORB23776-100
Anti-Adenovirus hexon Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
ELISA: 1:5-1:20

Type: Primary
Antigen: Adenovirus hexon
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR