Anti-C. trachomatis MOMP Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB23680-1EA 0 CHF
BIRBORB23680-1
Anti-C. trachomatis MOMP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: C. trachomatis MOMP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR