Anti-Mycobacterium avium Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB23677-0.2EA 0 CHF
BIRBORB23677-0.2
Anti-Mycobacterium avium Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Mycobacterium avium
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR