Anti-Influenza B (NP) Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB23404-1EA 801 CHF
BIRBORB23404-1
Anti-Influenza B (NP) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Influenza B (NP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR