Anti-RSV Fusion Protein Mouse Monoclonal Antibody

Lieferant: Biorbyt
BIRBORB23364-1EA 969 CHF
BIRBORB23364-1
Anti-RSV Fusion Protein Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RSV Fusion Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR