Anti-SARS-CoV, M Protein Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB23305-100EA 424 CHF
BIRBORB23305-100
Anti-SARS-CoV, M Protein Rabbit Polyclonal Antibody
Antibodies
ELISA: 10 ng

Type: Primary
Antigen: SARS-CoV, M Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR