Anti-Clostridium species Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB23240-1EA 774 CHF
BIRBORB23240-1
Anti-Clostridium species Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Clostridium species
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR