Anti-TMEFF2 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22915-5EA 259 CHF
BIRBORB22915-5 BIRBORB22915-20 BIRBORB22915-100
Anti-TMEFF2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TMEFF2 (transmembrane protein with EGF-like and two follistatin-like domains 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR