Anti-GST Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22847-500EA 0 CHF
BIRBORB22847-500 BIRBORB22847-1
Anti-GST Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR