Anti-SARS Spike Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22739-100EA 0 CHF
BIRBORB22739-100
Anti-SARS Spike Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SARS Spike
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR