Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB22703-1EA 0 CHF
BIRBORB22703-1 BIRBORB22703-500
Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a (Rat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR