Anti-B3GAT1 Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB22671-1EA 0 CHF
BIRBORB22671-1 BIRBORB22671-500
Anti-B3GAT1 Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR