Anti-CCR6 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22626-100EA 789 CHF
BIRBORB22626-100
Anti-CCR6 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CCR6 (chemokine (C-C motif) receptor 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR