Anti-IRF3 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22611-5EA 259 CHF
BIRBORB22611-5 BIRBORB22611-20 BIRBORB22611-100
Anti-IRF3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR