Anti-NQO1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22564-100EA 559 CHF
BIRBORB22564-100
Anti-NQO1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR