Anti-DBF4 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22513-100EA 0 CHF
BIRBORB22513-100
Anti-DBF4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DBF4 (DBF4 zinc finger)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR