Anti-MTHFD1L Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22509-100EA 559 CHF
BIRBORB22509-100
Anti-MTHFD1L Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: MTHFD1L (methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR